Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005493.2 | NP_005484.2 | 6 | 1058 | ||
XM_006714945.2 | XP_006715008.1 | 6 | 538 | |||
XM_011514205.2 | XP_011512507.1 | 6 | 1076 | |||
XM_017010149.1 | XP_016865638.1 | 6 | 446 | |||
GenBank mRNA | AB008515.1 | - | 6 | 747 | ||
AB055311.1 | - | 6 | 1058 | |||
AF306510.1 | - | 6 | 596 | |||
AK313334.1 | - | 5 | 449 | |||
BC019886.1 | - | 6 | 512 | |||
BC052781.1 | - | 6 | 1219 | |||
BC063849.1 | - | 4 | 314 | 6186252 | ||
KU921435.1 | - | 1 | 72 |