Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005493.2 | NP_005484.2 | 12 | 1934 | ||
XM_006714945.2 | XP_006715008.1 | 12 | 1414 | |||
XM_011514205.2 | XP_011512507.1 | 10 | 1682 | |||
XM_017010149.1 | XP_016865638.1 | 12 | 1322 | |||
GenBank mRNA | AB008515.1 | - | 12 | 1623 | ||
AB055311.1 | - | 12 | 1934 | |||
AF064606.1 | - | 10 | 947 | |||
AF306510.1 | - | 12 | 1472 | |||
AK313334.1 | - | 11 | 1325 | |||
BC019886.1 | - | 12 | 1388 | |||
BC052781.1 | - | 12 | 2095 | |||
BC063849.1 | - | 10 | 1190 | 6186252 | ||
KU921435.1 | - | 7 | 948 |
Gene Symbol: NOL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317724.1 | NP_001304653.1 | 9 | 881 | ||
GenBank mRNA | BU071935.1 | - | 9 | 634 |