Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005689.2 | NP_005680.1 | 12 | 2073 | ||
GenBank mRNA | AF070598.1 | - | 8 | 765 | ||
AF076775.1 | - | 12 | 1756 | |||
AF308472.1 | - | 12 | 2062 | |||
AJ289233.2 | - | 12 | 2033 | |||
AK026067.1 | - | 9 | 882 | |||
AK057026.1 | - | 12 | 1792 | |||
AK172812.1 | - | 12 | 1423 | |||
BC000559.2 | - | 12 | 2024 | 3162881 | ||
BC043423.1 | - | 5 | 2073 | |||
U66673.1 | - | 1 | 49 |