Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320724.1 | NP_001307653.1 | 5 | 893 | ||
NM_016940.2 | NP_058636.1 | 4 | 788 | |||
GenBank mRNA | AA406228.1 | - | 2 | 455 | ||
AF212232.1 | - | 4 | 749 | |||
AK000056.1 | - | 4 | 792 | |||
AK223357.1 | - | 4 | 799 | |||
BC012546.1 | - | 4 | 756 | 4291536 | ||
BC017912.1 | - | 4 | 774 | 4291966 | ||
BI710995.1 | - | 1 | 360 | |||
Y19009.1 | - | 4 | 767 |