Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005708.3 | NP_005699.1 | 2, 3 | 941 | ||
XM_011521044.2 | XP_011519346.1 | 2, 3 | 266 | |||
XM_017020298.1 | XP_016875787.1 | 2, 3 | 2549 | |||
XM_017020299.1 | XP_016875788.1 | 2, 3 | 1316 | |||
XM_017020300.1 | XP_016875789.1 | 2, 3 | 173 | |||
GenBank mRNA | AF105267.1 | - | 2, 3 | 894 | ||
AF111178.1 | - | 2, 3 | 943 | |||
AK074741.1 | - | 2, 3 | 395 | |||
AK130013.1 | - | 2, 3 | 278 | |||
AK290144.1 | - | 2, 3 | 462 | |||
AK304338.1 | - | 2, 3 | 760 | |||
AY358462.1 | - | 2, 3 | 874 | |||
BC106947.2 | - | 2, 3 | 789 | 40034564 |