Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005724.5 | NP_005715.1 | 2 | 364 | ||
NM_198902.2 | NP_944492.1 | 2 | 364 | |||
GenBank mRNA | AF054840.1 | - | 2 | 90 | ||
AF133423.1 | - | 2 | 164 | |||
AK001305.1 | - | 2 | 210 | |||
AK001310.1 | - | 2 | 196 | |||
AK001326.1 | - | 2 | 307 | |||
AK075336.1 | - | 2 | 245 | |||
AK075482.1 | - | 2 | 245 | |||
AK293677.1 | - | 2 | 182 | |||
BC000704.2 | - | 2 | 268 | 3349181 | ||
BC004280.1 | - | 2 | 226 | 3616380 | ||
BC009248.2 | - | 2 | 229 | 2964568 | ||
BC011206.1 | - | 2 | 258 | 3457158 | ||
DA359645.1 | - | 2 | 366 | |||
DB449004.1 | - | 2 | 310 |