Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005732.3 | NP_005723.2 | 5 | 1101 | ||
GenBank mRNA | AF057299.1 | - | 5 | 1021 | ||
AF057300.1 | - | 5 | 956 | |||
AK290597.1 | - | 5 | 1043 | |||
AK307439.1 | - | 5 | 776 | |||
BC062603.1 | - | 5 | 1018 | |||
BC073850.1 | - | 5 | 1018 | |||
BC108282.1 | - | 5 | 1038 | |||
BC136436.1 | - | 5 | 993 | 9020425 | ||
U63139.1 | - | 5 | 1088 |