Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005732.3 | NP_005723.2 | 8 | 1567 | ||
GenBank mRNA | AF057299.1 | - | 8 | 1487 | ||
AF057300.1 | - | 8 | 1422 | |||
AK290597.1 | - | 8 | 1509 | |||
AK307439.1 | - | 8 | 1242 | |||
BC062603.1 | - | 8 | 1484 | |||
BC073850.1 | - | 8 | 1484 | |||
BC108282.1 | - | 8 | 1504 | |||
BC136436.1 | - | 8 | 1459 | 9020425 | ||
U63139.1 | - | 8 | 1554 |