Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005735.3 | NP_005726.1 | 1 | 220 | ||
XM_005263854.4 | XP_005263911.1 | 1 | 55 | |||
GenBank mRNA | BC004374.2 | - | 1 | 88 | 3638252 | |
BC006372.2 | - | 1 | 53 | 4138683 | ||
BC010090.2 | - | 1 | 120 | 2989545 | ||
X82207.1 | - | 1 | 68 |