Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023037.2 | NP_075463.2 | 18 | 2506 | ||
XM_006719749.3 | XP_006719812.1 | 18 | 3587 | |||
XM_017020303.1 | XP_016875792.1 | 18 | 4064 | |||
XM_017020304.1 | XP_016875793.1 | 18 | 2506 | |||
XM_017020305.1 | XP_016875794.1 | 18 | 4083 | |||
GenBank mRNA | AL049784.1 | - | 18 | 2506 | ||
U50534.1 | - | 18 | 2505 |