Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282704.1 | NP_001269633.1 | 11, 12 | 1413 | ||
NM_001282705.1 | NP_001269634.1 | 10, 11 | 1435 | |||
NM_001282706.1 | NP_001269635.1 | 10, 11 | 1265 | |||
NM_001282707.1 | NP_001269636.1 | 10, 11 | 1240 | |||
NM_005742.3 | NP_005733.1 | 9, 10 | 1062 | |||
XM_011510308.1 | XP_011508610.1 | 9, 10 | 1003 | |||
GenBank mRNA | AK127433.1 | - | 11, 12 | 1413 | ||
AK131234.1 | - | 10, 11 | 1431 | |||
AK225496.1 | - | 8, 9 | 808 | |||
AK289428.1 | - | 9, 10 | 938 | |||
AK294347.1 | - | 10, 11 | 1155 | |||
AK297547.1 | - | 10, 11 | 1225 | |||
BC001312.1 | - | 9, 10 | 1018 | 3454007 | ||
D49489.1 | - | 1013 | ||||
U79278.1 | - | 8, 9 | 864 |