Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282704.1 | NP_001269633.1 | 7, 8 | 955 | ||
NM_001282705.1 | NP_001269634.1 | 6, 7 | 977 | |||
NM_001282706.1 | NP_001269635.1 | 6, 7 | 807 | |||
NM_001282707.1 | NP_001269636.1 | 6, 7 | 782 | |||
NM_005742.3 | NP_005733.1 | 5, 6 | 604 | |||
XM_011510308.1 | XP_011508610.1 | 5, 6 | 545 | |||
GenBank mRNA | AK127433.1 | - | 7, 8 | 955 | ||
AK131234.1 | - | 6, 7 | 973 | |||
AK225496.1 | - | 4, 5 | 350 | |||
AK289428.1 | - | 5, 6 | 480 | |||
AK294347.1 | - | 6, 7 | 697 | |||
AK297547.1 | - | 6, 7 | 767 | |||
BC001312.1 | - | 5, 6 | 560 | 3454007 | ||
BF979486.1 | - | 609 | ||||
D49489.1 | - | 555 | ||||
U79278.1 | - | 4, 5 | 406 |