Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARFRP1 SIRNA Assays ›
Gene Symbol:
ARFRP1
Gene Name:
ADP ribosylation factor related protein 1
Gene Aliases:
ARL18, ARP, Arp1
Chromosome Location:
Chr.20: 63698642 - 63708013 on Build GRCh38
UniGene ID:
Hs.389277
Species:
Human
siRNA ID
HSS115431
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARFRP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134758.3
NP_001128230.1
4
370
NM_001267544.2
NP_001254473.1
4
375
NM_001267545.2
NP_001254474.1
4
370
NM_001267546.2
NP_001254475.1
3
276
NM_001267547.2
NP_001254476.1
4
375
NM_001267548.2
NP_001254477.1
4
272
NM_001267549.2
NP_001254478.1
4
375
NM_003224.5
NP_003215.1
4
370
NR_051954.2
4
370
NR_051955.2
4
375
NR_051956.2
4
375
NR_051957.2
4
370
NR_051958.2
4
370
XM_011528482.2
XP_011526784.1
4
307
XM_011528483.1
XP_011526785.1
4
437
XM_017027576.1
XP_016883065.1
4
251
GenBank mRNA
AK095962.1
-
3
789
AL522492.3
-
3
227
AL533893.3
-
4
360
BC021023.1
-
4
255
BC092480.1
-
4
256
30531333
BC093967.1
-
4
273
7939812
BC143447.1
-
4
273
9051955
BC143448.1
-
4
278
9051956
BC143477.1
-
4
278
BI524032.1
-
4
369
BI819763.1
-
4
244
BM546222.1
-
295
BM549361.1
-
338
BT019624.1
-
3
222
BT019663.1
-
3
222
BU526941.1
-
4
312
BX342690.2
-
4
256
BX342805.2
-
4
349
BX463457.2
-
4
272
CA489361.1
-
3
214
CR456752.1
-
3
222
X91504.1
-
3
233
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
small GTPase mediated signal transduction
gastrulation
protein localization to Golgi apparatus
retrograde transport, endosome to Golgi
Golgi to plasma membrane protein transport
GTPase activity
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0