Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243877.1 | NP_001230806.1 | 3 | 861 | ||
NM_001243885.1 | NP_001230814.1 | 2 | 672 | |||
NM_005749.3 | NP_005740.1 | 2 | 875 | |||
GenBank mRNA | AA675891.1 | - | 2 | 586 | ||
AK313904.1 | - | 3 | 862 | |||
AY524047.1 | - | 1 | 442 | |||
BC031406.1 | - | 1 | 439 | 5194914 | ||
BC070493.1 | - | 2 | 863 | 6470511 | ||
BC098415.1 | - | 2 | 867 | 6480318 | ||
BT019380.1 | - | 1 | 432 | |||
BT019381.1 | - | 1 | 432 | |||
CR536487.1 | - | 1 | 432 | |||
CR541686.1 | - | 1 | 432 | |||
D38305.1 | - | 1 | 475 |