Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005752.4 | NP_005743.4 | 1 | 150 | ||
GenBank mRNA | AK310847.1 | - | 1 | 181 | ||
AK313316.1 | - | 1 | 150 | |||
AY358376.1 | - | 1 | 102 | |||
BC104414.2 | - | 1 | 139 | 40003144 | ||
BC104415.2 | - | 1 | 139 | 40003149 | ||
DA998903.1 | - | 1 | 150 |