Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017392.4 | NP_001017392.2 | 3 | 1652 | ||
NM_001321697.1 | NP_001308626.1 | 3 | 1808 | |||
NM_001321698.1 | NP_001308627.1 | 3 | 1600 | |||
NM_001321699.1 | NP_001308628.1 | 3 | 1600 | |||
NM_014884.4 | NP_055699.2 | 3 | 1652 | |||
XM_017026137.1 | XP_016881626.1 | 3 | 1808 | |||
GenBank mRNA | AB002363.2 | - | 3 | 1804 | ||
AF518874.1 | - | 3 | 1641 | |||
AK074842.1 | - | 3 | 1577 | |||
AK291275.1 | - | 3 | 1619 | |||
AK299860.1 | - | 3 | 1067 | |||
AL832488.1 | - | 3 | 1572 | |||
BC020586.1 | - | 3 | 1572 | 4400185 | ||
BX647813.1 | - | 3 | 1581 |