Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005755.2 | NP_005746.2 | 3 | 286 | ||
XM_011527619.2 | XP_011525921.1 | 4 | 535 | |||
GenBank mRNA | BC015364.1 | - | 3 | 331 | 3897136 | |
BC046112.1 | - | 3 | 285 | 5748376 | ||
L08187.1 | - | 3 | 291 |