Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005761.2 | NP_005752.1 | 22 | 3893 | ||
NR_037687.1 | 3 | 437 | ||||
XM_006719186.3 | XP_006719249.1 | 21 | 3881 | |||
XM_011537730.2 | XP_011536032.1 | 22 | 3920 | |||
XM_017018671.1 | XP_016874160.1 | 22 | 3920 | |||
XM_017018672.1 | XP_016874161.1 | 22 | 3920 | |||
GenBank mRNA | AB208934.1 | - | 21 | 2574 | ||
AF030339.1 | - | 22 | 3893 | |||
AK295226.1 | - | 7 | 1022 | |||
AK304327.1 | - | 3 | 437 | |||
AK307920.1 | - | 3 | 340 | |||
AK308985.1 | - | 8 | 826 |