Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005762.2 | NP_005753.1 | 13 | 2011 | ||
GenBank mRNA | AK131063.1 | - | 6 | 599 | ||
AK225760.1 | - | 13 | 2011 | |||
AK313478.1 | - | 13 | 1728 | |||
BC004978.1 | - | 13 | 2011 | 2906325 | ||
BC007390.2 | - | 2 | 328 | |||
BC052986.2 | - | 11 | 1762 | 6502540 | ||
U31657.1 | - | 2 | 267 | |||
U78773.1 | - | 13 | 1822 | |||
U95040.1 | - | 13 | 1978 | |||
X97548.1 | - | 13 | 2083 |