Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005763.3 | NP_005754.2 | 17 | 1964 | ||
XM_011515725.2 | XP_011514027.1 | 17 | 1943 | |||
GenBank mRNA | AF229180.1 | - | 17 | 1888 | ||
AJ007714.2 | - | 16 | 1915 | |||
AK225898.1 | - | 17 | 1857 | |||
AK294326.1 | - | 6 | 514 | |||
BC144247.1 | - | 16 | 1874 |