Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014252.3 | NP_055067.1 | 5 | 842 | ||
GenBank mRNA | AF112968.1 | - | 5 | 679 | ||
AF177333.1 | - | 5 | 644 | |||
AK296862.1 | - | 6 | 494 | |||
AK313602.1 | - | 5 | 724 | |||
AL538057.3 | - | 5 | 834 | |||
BC002702.2 | - | 5 | 756 | 3614008 | ||
BU726339.1 | - | 3 | 560 |
Gene Symbol: TPTE2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093232.1 | - | 1 | 670 |