Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005778.3 | NP_005769.1 | 17, 18 | 1629 | ||
NR_036627.2 | 16, 17 | 1555 | ||||
XM_011533261.1 | XP_011531563.1 | 15, 16 | 1407 | |||
XM_011533262.1 | XP_011531564.1 | 17, 18 | 1629 | |||
XM_017005503.1 | XP_016860992.1 | 17, 18 | 1611 | |||
XM_017005505.1 | XP_016860994.1 | 16, 17 | 1522 | |||
XM_017005506.1 | XP_016860995.1 | 14, 15 | 1372 | |||
GenBank mRNA | AB208813.1 | - | 17, 18 | 4115 | ||
AF091263.1 | - | 17, 18 | 1602 | |||
AF103802.4 | - | 17, 18 | 1580 | |||
AK297249.1 | - | 16, 17 | 1509 | |||
AK297445.1 | - | 17, 18 | 1484 | |||
AK302766.1 | - | 16, 17 | 1437 | |||
AK314032.1 | - | 17, 18 | 1584 | |||
BC002957.1 | - | 17, 18 | 1575 | |||
BX441555.2 | - | 3, 4 | 271 | |||
KU178505.1 | - | 16, 17 | 1454 | |||
KU178506.1 | - | 15, 16 | 1380 | |||
U23946.1 | - | 17, 18 | 1523 |