Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242826.1 | NP_001229755.1 | 6 | 887 | ||
NM_005785.3 | NP_005776.1 | 6 | 948 | |||
NM_194358.2 | NP_919339.1 | 7 | 1091 | |||
NM_194359.2 | NP_919340.1 | 6 | 843 | |||
NR_040053.1 | 7 | 1091 | ||||
XM_005268561.4 | XP_005268618.1 | 7 | 999 | |||
XM_011537734.2 | XP_011536036.1 | 7 | 1005 | |||
XM_011537735.2 | XP_011536037.1 | 7 | 922 | |||
GenBank mRNA | AF077599.1 | - | 5 | 781 | ||
AK300752.1 | - | 7 | 949 | |||
AK304576.1 | - | 7 | 788 | |||
AK314811.1 | - | 6 | 807 | |||
AY007109.1 | - | 1 | 66 | |||
BC024284.1 | - | 7 | 1069 | |||
BC032637.1 | - | 5 | 796 | 5496849 | ||
BC036267.1 | - | 6 | 892 | |||
BC095418.1 | - | 7 | 1026 |