Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308210.1 | NP_001295139.1 | 2 | 601 | ||
NM_005786.5 | NP_005777.3 | 2 | 702 | |||
XM_005266641.3 | XP_005266698.1 | 2 | 972 | |||
GenBank mRNA | AB210042.1 | - | 1 | 2797 | ||
AK127042.1 | - | 1 | 1443 | |||
AL833321.1 | - | 2 | 270 | |||
BX447393.2 | - | 2 | 270 |