Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005791.2 | NP_005782.1 | 9 | 1979 | ||
GenBank mRNA | AK056862.1 | - | 9 | 1979 | ||
BC041084.1 | - | 9 | 1629 | |||
BC126389.1 | - | 9 | 1916 | 8992105 | ||
X98494.1 | - | 9 | 1602 |