Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PATJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176877.2 | NP_795352.2 | 31 | 4200 | ||
XM_005270341.3 | XP_005270398.1 | 31 | 4198 | |||
XM_011540462.2 | XP_011538764.1 | 31 | 4198 | |||
XM_011540463.2 | XP_011538765.1 | 31 | 4386 | |||
XM_011540464.2 | XP_011538766.1 | 31 | 4198 | |||
XM_011540465.2 | XP_011538767.1 | 31 | 4198 | |||
XM_011540466.2 | XP_011538768.1 | 31 | 4198 | |||
XM_011540467.2 | XP_011538769.1 | 31 | 4198 | |||
XM_011540468.2 | XP_011538770.1 | 31 | 4198 | |||
XM_016999998.1 | XP_016855487.1 | 31 | 4198 | |||
XM_016999999.1 | XP_016855488.1 | 31 | 4198 | |||
XM_017000000.1 | XP_016855489.1 | 26 | 3444 | |||
XM_017000001.1 | XP_016855490.1 | 12 | 1275 | |||
GenBank mRNA | AF397170.1 | - | 30 | 4086 | ||
AK130492.1 | - | 11 | 1162 | |||
AK308665.1 | - | 5 | 763 | |||
BC144135.1 | - | 31 | 4152 |