Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318875.1 | NP_001305804.1 | 6, 7 | 773 | ||
NM_005803.3 | NP_005794.1 | 7, 8 | 917 | |||
XM_005248780.3 | XP_005248837.1 | 7, 8 | 650 | |||
XM_006714947.3 | XP_006715010.1 | 6, 7 | 754 | |||
XM_017010157.1 | XP_016865646.1 | 7, 8 | 571 | |||
XM_017010158.1 | XP_016865647.1 | 5, 6 | 472 | |||
GenBank mRNA | AF085357.1 | - | 7, 8 | 706 | ||
AF089750.2 | - | 7, 8 | 742 | |||
AF117234.1 | - | 7, 8 | 743 | |||
AK000816.1 | - | 7, 8 | 753 | |||
AK222530.1 | - | 7, 8 | 753 | |||
AK301291.1 | - | 6, 7 | 607 | |||
BC001146.1 | - | 6, 7 | 600 | 3534771 | ||
BT007077.1 | - | 6, 7 | 578 | |||
CR456946.1 | - | 6, 7 | 578 |