Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190983.1 | NP_001177912.1 | 3 | 351 | ||
NM_016138.4 | NP_057222.2 | 3 | 402 | |||
GenBank mRNA | AF032900.1 | - | 2 | 218 | ||
AF098948.1 | - | 3 | 360 | |||
AF374413.1 | - | 2 | 291 | |||
AK024291.1 | - | 3 | 383 | |||
AK225115.1 | - | 3 | 351 | |||
AK315470.1 | - | 3 | 402 | |||
AL136647.1 | - | 3 | 354 | |||
BC003185.1 | - | 3 | 393 | 3503763 | ||
BC043272.1 | - | 3 | 1152 | 5297074 | ||
U81276.1 | - | 1 | 82 |