Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005822.3 | NP_005813.2 | 1 | 474 | ||
GenBank mRNA | AK312392.1 | - | 1 | 181 | ||
BC038509.1 | - | 1 | 257 | 5259147 | ||
D83407.1 | - | 1 | 250 | |||
DA106611.1 | - | 1 | 474 | |||
DA207634.1 | - | 1 | 463 |