Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278784.1 | NP_001265713.1 | 3 | 356 | ||
NM_005827.2 | NP_005818.2 | 3 | 381 | |||
XM_005256929.3 | XP_005256986.1 | 4 | 296 | |||
XM_006721632.3 | XP_006721695.1 | 4 | 500 | |||
XM_011524179.2 | XP_011522481.1 | 2 | 266 | |||
XM_011524180.2 | XP_011522482.1 | 3 | 232 | |||
GenBank mRNA | AK098056.1 | - | 3 | 563 | ||
AK293563.1 | - | 3 | 381 | |||
AK296073.1 | - | 3 | 350 | |||
AU309730.1 | - | 3 | 390 | |||
BC011888.2 | - | 3 | 360 | 3832062 | ||
BX641166.1 | - | 4 | 328 | |||
D87989.1 | - | 3 | 350 | |||
Z36793.1 | - | 2 | 137 |