Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCOCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261390.1 | NP_001248319.1 | 2 | 245 | ||
NM_001261391.1 | NP_001248320.1 | 2 | 245 | |||
NM_001261393.1 | NP_001248322.1 | 2 | 245 | |||
NM_005831.4 | NP_005822.1 | 2 | 245 | |||
GenBank mRNA | AK222666.1 | - | 2 | 195 | ||
AK223227.1 | - | 2 | 188 | |||
AK293329.1 | - | 2 | 207 | |||
AK298177.1 | - | 2 | 188 | |||
AK303313.1 | - | 2 | 198 | |||
AK304024.1 | - | 2 | 198 | |||
AK310648.1 | - | 2 | 198 | |||
AK314796.1 | - | 2 | 198 | |||
AL046722.1 | - | 2 | 245 | |||
BC004130.2 | - | 2 | 184 | 3505988 | ||
BC015893.2 | - | 1 | 154 | 3921397 | ||
BG773301.1 | - | 2 | 227 | |||
CR456763.1 | - | 1 | 144 | |||
U22897.1 | - | 2 | 198 |