Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144823.2 | NP_001138295.1 | 23 | 3634 | ||
NM_001320835.1 | NP_001307764.1 | 23 | 3637 | |||
NM_005848.4 | NP_005839.3 | 22 | 3498 | |||
XM_005254120.4 | XP_005254177.1 | 22 | 3416 | |||
XM_005254121.3 | XP_005254178.1 | 22 | 3505 | |||
XM_005254122.4 | XP_005254179.1 | 23 | 3619 | |||
XM_006720371.3 | XP_006720434.1 | 23 | 3372 | |||
XM_011521156.2 | XP_011519458.1 | 23 | 3632 | |||
XM_011521157.2 | XP_011519459.1 | 13 | 1972 | |||
XM_017021862.1 | XP_016877351.1 | 22 | 3357 | |||
XM_017021863.1 | XP_016877352.1 | 21 | 3283 | |||
XM_017021864.1 | XP_016877353.1 | 23 | 3632 | |||
XM_017021865.1 | XP_016877354.1 | 22 | 3486 | |||
GenBank mRNA | AF534403.1 | - | 22 | 3466 | ||
AK091268.1 | - | 2 | 214 | |||
AL832602.2 | - | 20 | 3024 | |||
AL833317.1 | - | 23 | 3465 | |||
AL833384.1 | - | 19 | 2769 | |||
BC041706.1 | - | 22 | 3329 | 5530033 |