Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005877.5 | NP_005868.1 | 11 | 1767 | ||
GenBank mRNA | AK098463.1 | - | 4 | 533 | ||
AK303273.1 | - | 12 | 1692 | |||
BC001976.1 | - | 11 | 1707 | 3459964 | ||
BC007684.2 | - | 11 | 1734 | 3634035 | ||
CR456575.1 | - | 11 | 1710 | |||
X85237.1 | - | 11 | 1709 |