Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270660.1 | NP_001257589.1 | 9 | 1001 | ||
NM_001270661.1 | NP_001257590.1 | 7 | 830 | |||
NM_005885.3 | NP_005876.2 | 10 | 1145 | |||
XM_011513932.2 | XP_011512234.1 | 10 | 903 | |||
XM_011513934.1 | XP_011512236.1 | 10 | 1106 | |||
XM_011513936.2 | XP_011512238.1 | 10 | 1106 | |||
XM_017008944.1 | XP_016864433.1 | 10 | 1106 | |||
GenBank mRNA | AB011169.1 | - | 10 | 1068 | ||
AK000970.1 | - | 10 | 1060 | |||
AK296585.1 | - | 7 | 753 | |||
AK299339.1 | - | 6 | 483 | |||
AK300034.1 | - | 9 | 937 | |||
BC046148.1 | - | 2 | 62 | |||
BC136461.1 | - | 10 | 1017 | 9020450 | ||
BC136462.1 | - | 10 | 1017 | 9020451 | ||
BC142679.1 | - | 10 | 996 | 40148250 | ||
BC142694.1 | - | 10 | 996 | 40148713 |