Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270660.1 | NP_001257589.1 | 23 | 2516 | ||
NM_001270661.1 | NP_001257590.1 | 21 | 2345 | |||
NM_005885.3 | NP_005876.2 | 24 | 2660 | |||
XM_011513932.2 | XP_011512234.1 | 24 | 2418 | |||
XM_011513937.2 | XP_011512239.1 | 14 | 2002 | |||
GenBank mRNA | AB011169.1 | - | 24 | 2583 | ||
AF009301.1 | - | 11 | 1279 | |||
AK296585.1 | - | 21 | 2268 | |||
AK300034.1 | - | 23 | 2452 | |||
BC046148.1 | - | 16 | 1577 | |||
BC136461.1 | - | 24 | 2532 | 9020450 | ||
BC136462.1 | - | 24 | 2532 | 9020451 | ||
BC142679.1 | - | 24 | 2511 | 40148250 | ||
BC142694.1 | - | 24 | 2511 | 40148713 |