Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265588.1 | NP_001252517.1 | 5 | 1353 | ||
NM_003414.5 | NP_003405.3 | 4 | 1202 | |||
NR_049749.1 | 5 | 1246 | ||||
XM_017022865.1 | XP_016878354.1 | 5 | 1289 | |||
XM_017022866.1 | XP_016878355.1 | 6 | 1273 | |||
GenBank mRNA | AF220492.1 | - | 4 | 1155 | ||
AK292573.1 | - | 4 | 1027 | |||
AL832123.1 | - | 1 | 3389 | |||
BC036367.1 | - | 4 | 1126 | 4828269 | ||
BC127088.1 | - | 4 | 1125 | 40123276 | ||
BC127089.1 | - | 4 | 1125 | 40123277 | ||
X78925.1 | - | 3 | 960 |