Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202413.1 | NP_001189342.1 | 5 | 624 | ||
NM_001202414.1 | NP_001189343.1 | 6 | 752 | |||
NM_006066.3 | NP_006057.1 | 5 | 841 | |||
NM_153326.2 | NP_697021.1 | 4 | 713 | |||
XM_011540491.2 | XP_011538793.1 | 3 | 572 | |||
XM_011540492.2 | XP_011538794.1 | 4 | 538 | |||
GenBank mRNA | AK293083.1 | - | 4 | 449 | ||
AK307068.1 | - | 4 | 543 | |||
AK307237.1 | - | 4 | 636 | |||
AK307288.1 | - | 4 | 606 | |||
AK311175.1 | - | 4 | 473 | |||
AU142853.1 | - | 5 | 563 | |||
BC000670.2 | - | 5 | 770 | 3349300 | ||
BC005394.1 | - | 4 | 429 | 4051944 | ||
BT007003.1 | - | 3 | 334 | |||
CR457010.1 | - | 3 | 334 | |||
CX788871.1 | - | 6 | 679 | |||
EU668319.1 | - | 4 | 670 | |||
EU794588.1 | - | 4 | 670 | |||
J04794.1 | - | 4 | 394 |