Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142288.1 | NP_001135760.1 | 2 | 500 | ||
NM_006067.4 | NP_006058.1 | 2 | 500 | |||
XM_011522813.2 | XP_011521115.1 | 2 | 702 | |||
XM_017022867.1 | XP_016878356.1 | 2 | 191 | |||
GenBank mRNA | AF005888.1 | - | 2 | 369 | ||
AK313977.1 | - | 2 | 482 | |||
BC001472.2 | - | 2 | 452 | 3138396 | ||
BC005886.2 | - | 2 | 452 | 3546377 | ||
BC007445.1 | - | 2 | 443 | 3009805 | ||
BC020250.1 | - | 2 | 452 | 4653699 | ||
BE855452.1 | - | 2 | 374 | |||
BP307869.1 | - | 2 | 505 | |||
BT007132.1 | - | 2 | 253 |