Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006067.4 | NP_006058.1 | 4, 5 | 714 | ||
XM_017022867.1 | XP_016878356.1 | 4, 5 | 405 | |||
GenBank mRNA | AF005888.1 | - | 4, 5 | 583 | ||
AK313977.1 | - | 4, 5 | 696 | |||
BC001472.2 | - | 4, 5 | 666 | 3138396 | ||
BC005886.2 | - | 4, 5 | 666 | 3546377 | ||
BC007445.1 | - | 4, 5 | 657 | 3009805 | ||
BC020250.1 | - | 4, 5 | 666 | 4653699 | ||
BT007132.1 | - | 4, 5 | 467 |