Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006068.4 | NP_006059.2 | 2 | 2343 | ||
XM_005262637.4 | XP_005262694.1 | 2 | 2328 | |||
XM_011513612.2 | XP_011511914.1 | 3 | 2496 | |||
XM_011513613.2 | XP_011511915.1 | 2 | 2729 | |||
XM_011513614.2 | XP_011511916.1 | 2 | 2366 | |||
GenBank mRNA | AB020807.1 | - | 1 | 2289 | ||
AB445652.1 | - | 1 | 2222 | |||
AK313618.1 | - | 2 | 2344 | |||
BC111755.1 | - | 2 | 1300 | 40007924 |