Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317836.1 | NP_001304765.1 | 6, 7 | 733 | ||
NM_006315.5 | NP_006306.2 | 5, 6 | 634 | |||
GenBank mRNA | AK057124.1 | - | 5, 6 | 1571 | ||
AK125801.1 | - | 6, 7 | 1578 | |||
AK225892.1 | - | 5, 6 | 633 | |||
AK300290.1 | - | 6, 7 | 642 | |||
AU132917.1 | - | 5, 6 | 591 | |||
BC107061.2 | - | 3, 4 | 259 | 40006331 | ||
BQ882065.1 | - | 5, 6 | 477 | |||
BX648409.1 | - | 1, 2 | 982 | |||
CA454695.1 | - | 29 |