Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288985.1 | NP_001275914.1 | 17, 18 | 2475 | ||
NM_001288986.1 | NP_001275915.1 | 16, 17 | 2314 | |||
NM_007168.3 | NP_009099.1 | 15, 16 | 2194 | |||
XM_005256938.2 | XP_005256995.1 | 16, 17 | 2342 | |||
XM_005256940.4 | XP_005256997.1 | 15, 16 | 2247 | |||
XM_011524191.2 | XP_011522493.1 | 16, 17 | 2305 | |||
XM_011524192.2 | XP_011522494.1 | 16, 17 | 2305 | |||
GenBank mRNA | AB020629.1 | - | 15, 16 | 2153 | ||
AK295876.1 | - | 16, 17 | 2287 | |||
AK304286.1 | - | 15, 16 | 2146 | |||
AK304360.1 | - | 16, 17 | 2285 | |||
BC037309.1 | - | 17, 18 | 2242 | |||
BC130280.1 | - | 17, 18 | 2324 | 40146311 |