Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286149.1 | NP_001273078.1 | 7 | 795 | ||
NM_001286150.1 | NP_001273079.1 | 7 | 815 | |||
NM_001286151.1 | NP_001273080.1 | 7 | 837 | |||
NM_006085.5 | NP_006076.4 | 8 | 923 | |||
XM_005272998.2 | XP_005273055.1 | 9 | 939 | |||
XM_005272999.4 | XP_005273056.1 | 9 | 1048 | |||
XM_005273000.4 | XP_005273057.1 | 8 | 972 | |||
XM_005273004.2 | XP_005273061.1 | 7 | 696 | |||
XM_005273005.4 | XP_005273062.1 | 7 | 808 | |||
XM_006711114.3 | XP_006711177.1 | 8 | 912 | |||
XM_011509063.2 | XP_011507365.1 | 8 | 886 | |||
XM_011509064.2 | XP_011507366.1 | 7 | 795 | |||
XM_017000043.1 | XP_016855532.1 | 7 | 887 | |||
XM_017000044.1 | XP_016855533.1 | 6 | 722 | |||
XM_017000046.1 | XP_016855535.1 | 6 | 646 | |||
GenBank mRNA | AF125042.1 | - | 8 | 860 | ||
AJ249339.1 | - | 8 | 888 | |||
AK021941.1 | - | 3 | 321 | |||
AK291943.1 | - | 8 | 858 | |||
AK298476.1 | - | 7 | 740 | |||
AK300777.1 | - | 7 | 760 | |||
AK315962.1 | - | 7 | 817 | |||
BC017801.1 | - | 8 | 862 | 4338207 | ||
EU794594.1 | - | 8 | 860 |