Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006089.2 | NP_006080.1 | 14, 15 | 2122 | ||
NR_033717.1 | 14, 15 | 2122 | ||||
XM_017029219.1 | XP_016884708.1 | 14, 15 | 2018 | |||
XM_017029220.1 | XP_016884709.1 | 14, 15 | 2014 | |||
XM_017029222.1 | XP_016884711.1 | 8, 9 | 1400 | |||
GenBank mRNA | AL833937.1 | - | 2, 3 | 407 | ||
BC051913.1 | - | 14, 15 | 2064 | |||
BC064617.1 | - | 14, 15 | 2079 | 5575894 | ||
Y18004.1 | - | 14, 15 | 2056 |