Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190456.1 | NP_001177385.1 | 2, 3 | 282 | ||
NM_001190457.1 | NP_001177386.1 | 2, 3 | 561 | |||
NM_001324014.1 | NP_001310943.1 | 3, 4 | 493 | |||
NM_001324015.1 | NP_001310944.1 | 2, 3 | 294 | |||
NM_006091.4 | NP_006082.3 | 2, 3 | 516 | |||
GenBank mRNA | AB010098.1 | - | 2, 3 | 249 | ||
AB023142.1 | - | 1, 2 | 220 | |||
AK289678.1 | - | 2, 3 | 561 | |||
BC026335.1 | - | 2, 3 | 232 | 4792415 | ||
BX461395.2 | - | 2, 3 | 515 | |||
DA128408.1 | - | 2, 3 | 282 | |||
DA182763.1 | - | 2, 3 | 251 | |||
DA219458.1 | - | 2, 3 | 243 | |||
DA291058.1 | - | 2, 3 | 243 |