Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANAPC10 SIRNA Assays ›
Gene Symbol:
ANAPC10
Gene Name:
anaphase promoting complex subunit 10
Gene Aliases:
APC10, DOC1
Chromosome Location:
Chr.4: 144967117 - 145098541 on Build GRCh38
UniGene ID:
Hs.480876
Species:
Human
siRNA ID
HSS115911
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANAPC10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256706.1
NP_001243635.1
3
284
NM_001256707.1
NP_001243636.1
4
357
NM_001256708.1
NP_001243637.1
3
490
NM_001256709.1
NP_001243638.1
5
425
NM_001256710.1
NP_001243639.1
3
284
NM_001256711.1
NP_001243640.1
4
357
NM_001256712.1
NP_001243641.1
3
273
NM_001318367.1
NP_001305296.1
3
322
NM_014885.4
NP_055700.2
3
273
NR_046345.1
2
157
XM_011531524.2
XP_011529826.1
4
513
XM_011531525.2
XP_011529827.1
3
300
XM_011531526.2
XP_011529828.1
3
300
XM_011531527.2
XP_011529829.1
3
300
XM_011531529.2
XP_011529831.1
3
300
XM_011531530.2
XP_011529832.1
4
316
XM_011531531.2
XP_011529833.1
3
300
XM_011531532.2
XP_011529834.1
3
274
XM_017007638.1
XP_016863127.1
3
300
XM_017007639.1
XP_016863128.1
4
949
XM_017007640.1
XP_016863129.1
3
341
XM_017007641.1
XP_016863130.1
4
769
XM_017007642.1
XP_016863131.1
4
514
XM_017007643.1
XP_016863132.1
3
300
XM_017007644.1
XP_016863133.1
3
274
GenBank mRNA
AB012109.1
-
3
271
AF132794.1
-
3
228
AL080090.1
-
4
331
AV757106.2
-
3
332
BC005217.1
-
3
250
3929766
BF693880.1
-
3
282
BG494966.1
-
5
387
BG530912.1
-
4
328
BI488888.1
-
2
155
BI913723.1
-
3
323
BU175327.1
-
3
254
BU567906.1
-
3
493
CN305215.1
-
2
152
CV023091.1
-
2
172
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
mitotic nuclear division
negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
regulation of mitotic metaphase/anaphase transition
anaphase-promoting complex-dependent catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
cell division
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
protein K11-linked ubiquitination
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline