Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098531.2 | NP_001092001.1 | 21, 22 | 2792 | ||
NM_001098532.2 | NP_001092002.1 | 20, 21 | 2556 | |||
NM_006105.5 | NP_006096.2 | 21, 22 | 2326 | |||
XM_005268571.3 | XP_005268628.1 | 20, 21 | 2302 | |||
XM_011537752.1 | XP_011536054.1 | 21, 22 | 2809 | |||
XM_011537755.1 | XP_011536057.1 | 21, 22 | 2371 | |||
XM_011537758.2 | XP_011536060.1 | 21, 22 | 2421 | |||
GenBank mRNA | AF103905.1 | - | 20, 21 | 2116 | ||
AK055502.1 | - | 3, 4 | 1107 | |||
AK074444.1 | - | 5, 6 | 1484 | |||
AK092448.1 | - | 21, 22 | 2764 | |||
AK125545.1 | - | 17, 18 | 1809 | |||
AK290230.1 | - | 22, 23 | 2457 | |||
BC017728.1 | - | 21, 22 | 2406 | 4414360 | ||
BC092404.1 | - | 21, 22 | 2395 | 6181000 |
Gene Symbol: LOC105369748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL831948.1 | - | 4 | 1053 |