Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039619.2 | NP_001034708.1 | 3, 4 | 632 | ||
NM_001282954.1 | NP_001269883.1 | 2, 3 | 513 | |||
NM_006109.4 | NP_006100.2 | 3, 4 | 547 | |||
XM_017020927.1 | XP_016876416.1 | 2, 3 | 271 | |||
GenBank mRNA | AB451246.1 | - | 3, 4 | 323 | ||
AB451370.1 | - | 3, 4 | 323 | |||
AF015913.1 | - | 3, 4 | 323 | |||
AF167572.1 | - | 3, 4 | 414 | |||
AK075251.1 | - | 3, 4 | 425 | |||
AK300863.1 | - | 2, 3 | 297 | |||
AK315500.1 | - | 3, 4 | 363 | |||
AL833612.1 | - | 3, 4 | 332 | |||
BC025979.2 | - | 3, 4 | 335 | 3833019 | ||
CR456741.1 | - | 3, 4 | 323 |