Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320049.1 | NP_001306978.1 | 5 | 640 | ||
NM_006331.7 | NP_006322.4 | 6 | 790 | |||
GenBank mRNA | AI394112.1 | - | 1 | 279 | ||
AK291597.1 | - | 6 | 655 | |||
AK307177.1 | - | 3 | 423 | |||
BC055314.1 | - | 6 | 682 | 4718702 | ||
BP315329.1 | - | 5 | 508 | |||
BQ015307.1 | - | 1 | 275 | |||
U72514.1 | - | 6 | 633 |