Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006336.3 | NP_006327.2 | 6 | 1472 | ||
XM_005251645.2 | XP_005251702.1 | 6 | 1264 | |||
XM_005251646.2 | XP_005251703.1 | 6 | 1467 | |||
XM_011518122.2 | XP_011516424.1 | 6 | 1274 | |||
XM_017014185.1 | XP_016869674.1 | 6 | 1880 | |||
XM_017014186.1 | XP_016869675.1 | 6 | 1261 | |||
XM_017014187.1 | XP_016869676.1 | 6 | 1880 | |||
XM_017014188.1 | XP_016869677.1 | 6 | 1248 | |||
XM_017014189.1 | XP_016869678.1 | 6 | 1467 | |||
XM_017014190.1 | XP_016869679.1 | 6 | 1261 | |||
GenBank mRNA | AK314911.1 | - | 6 | 1220 | ||
BC052563.1 | - | 6 | 1457 | 6303965 | ||
X99802.1 | - | 5 | 1062 |